Learn More
Medchemexpress LLC HY-111905 5mg Medchemexpress, BRD7-IN-1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1119055MG
Encompass_Preferred
Medchemexpress, HY-111905 5mg BRD7-IN-1 CAS: BRD7-IN-1, a modified derivative of BI7273 (BRD7/9 inhibitor), binds to a VHL ligand via a linker to form a PROTAC VZ185 (VZ185 against BRD7/9 with DC 50 s of 4.5 and 1.8 nM, respectively) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2405