Learn More
Medchemexpress LLC HY-12185 10mg Medchemexpress, BRD7389 CAS:376382-11-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1218510MG
Encompass_Preferred
Medchemexpress, HY-12185 10mg BRD7389 CAS:376382-11-5 BRD7389 is a specific RSK family kinase inhibitor with IC50s of 1.5 μM, 2.4 μM, and 1.2 μM for RSK1, RSK2, and RSK3, respectively. BRD7389 is a small-molecule inducer of insulin expression in pancreatic α-cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9626