Learn More
TARGETMOL CHEMICALS INC BRD73954 10MG
SDP
Catalog No. 502579384
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BRD73954 an effective and specific HDAC inhibitor which is with IC50 of 36 nM and 120 nM for HDAC6 and HDAC8 respectively. purity: 99%
Catalog No. 50-257-9384 Supplier TARGETMOL CHEMICALS INC Supplier No. T642110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BRD73954 an effective and specific HDAC inhibitor which is with IC50 of 36 nM and 120 nM for HDAC6 and HDAC8 respectively. purity: 99%