Learn More
Medchemexpress LLC BRD73954 | 1440209-96-0 | 99.1% | 284.31 | 250 MG
SDP
Catalog No. 5000384196
Encompass_Preferred
BRD73954 is a potent HDAC inhibitor that selectively inhibits HDAC6 and HDAC8 with IC50 values of 0.0036 μM and 0.12 μM, respectively. It also inhibits HDAC2, HDAC1, and HDAC3 at higher concentrations. It leads to decreased levels of HDAC6 and upregulation of Ac-Tubulin.
  • Potent HDAC inhibitor
  • Selectively inhibits HDAC6 (IC50 = 0.0036 μM) and HDAC8 (IC50 = 0.12 μM)
  • Also inhibits HDAC2 (IC50 = 9 μM), HDAC1 (IC50 = 12 μM), and HDAC3 (IC50 = 23 μM)
  • Decreases HDAC6 levels
  • Associated with upregulation of Ac-Tubulin
  • Available in solid and solution forms
Catalog No. 50-003-84196 Supplier Medchemexpress LLC Supplier No. 250MGHY18700
BRD73954 is a potent HDAC inhibitor that selectively inhibits HDAC6 and HDAC8 with IC50 values of 0.0036 μM and 0.12 μM, respectively. It also inhibits HDAC2, HDAC1, and HDAC3 at higher concentrations. It leads to decreased levels of HDAC6 and upregulation of Ac-Tubulin.
  • Potent HDAC inhibitor
  • Selectively inhibits HDAC6 (IC50 = 0.0036 μM) and HDAC8 (IC50 = 0.12 μM)
  • Also inhibits HDAC2 (IC50 = 9 μM), HDAC1 (IC50 = 12 μM), and HDAC3 (IC50 = 23 μM)
  • Decreases HDAC6 levels
  • Associated with upregulation of Ac-Tubulin
  • Available in solid and solution forms