Learn More
Medchemexpress LLC HY-19694 5mg Medchemexpress, BRD7552 CAS:1137359-47-7 Purity:>98%
SDP
Catalog No. 502082162
Encompass_Preferred
Medchemexpress, HY-19694 5mg BRD7552 CAS:1137359-47-7 BRD7552, a potent PDX1 transcription factor inducer, upregulates PDX1 expression in both primary human islets and ductal cells, and induces epigenetic changes in the PDX1 promoter consistent with transcriptional activation. BRD7552 increases insulin expression. PDX1 is a key transcription factor involved in pancreas development and β cell function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2162 Supplier Medchemexpress LLC Supplier No. HY196945MG
Medchemexpress, HY-19694 5mg BRD7552 CAS:1137359-47-7 BRD7552, a potent PDX1 transcription factor inducer, upregulates PDX1 expression in both primary human islets and ductal cells, and induces epigenetic changes in the PDX1 promoter consistent with transcriptional activation. BRD7552 increases insulin expression. PDX1 is a key transcription factor involved in pancreas development and β cell function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.