Learn More
Aviva Systems Biology Corp BRD9 Antibody - N-terminal region (ARP34832_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34832_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BRD9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BRD9. Alternate names: PRO9856, LAVS3040ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55580