Learn More
Biomatik Corporation Brd9 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10597
Encompass_Preferred
Brd9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to BRD9. The Brd9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Bromodomain-Containing Protein 9 Protein (338-597Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: BRD9. Accession Number: Q9H8M2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7495