Learn More
Medchemexpress LLC Brd9500 | 1630760-75-6 | 98.4% | 291.32 | 100 MG
SDP
Catalog No. 5000352212
Encompass_Preferred
BRD9500 is an orally active phosphodiesterases 3 (PDE3) inhibitor, functioning as a DNMDP analog. It potently and selectively inhibits PDE3A (IC50 = 10 nM) and PDE3B (IC50 = 27 nM), exhibiting antitumor activity by inducing PDE3A/B interactions with SFLN12. It shows an EC50 of 1 nM for SK-MEL-3 melanoma cell line viability and an EC50 of 1.6 nM for HeLa cell viability. In vivo studies demonstrate that BRD9500 inhibits SK-MEL-3 melanoma growth in mice and achieves high plasma levels, making it a valuable candidate for xenograft testing.
  • Orally active phosphodiesterases 3 (PDE3) inhibitor
  • Potent and selective inhibition of PDE3A and PDE3B
  • Antitumor activity, including inhibition of SK-MEL-3 melanoma growth in mice
  • Kills cancer cells by inducing PDE3A/B interactions with SFLN12
  • Demonstrates high plasma levels in mice, suitable for in vivo xenograft testing
Catalog No. 50-003-52212 Supplier Medchemexpress LLC Supplier No. HY136350100MG
BRD9500 is an orally active phosphodiesterases 3 (PDE3) inhibitor, functioning as a DNMDP analog. It potently and selectively inhibits PDE3A (IC50 = 10 nM) and PDE3B (IC50 = 27 nM), exhibiting antitumor activity by inducing PDE3A/B interactions with SFLN12. It shows an EC50 of 1 nM for SK-MEL-3 melanoma cell line viability and an EC50 of 1.6 nM for HeLa cell viability. In vivo studies demonstrate that BRD9500 inhibits SK-MEL-3 melanoma growth in mice and achieves high plasma levels, making it a valuable candidate for xenograft testing.
  • Orally active phosphodiesterases 3 (PDE3) inhibitor
  • Potent and selective inhibition of PDE3A and PDE3B
  • Antitumor activity, including inhibition of SK-MEL-3 melanoma growth in mice
  • Kills cancer cells by inducing PDE3A/B interactions with SFLN12
  • Demonstrates high plasma levels in mice, suitable for in vivo xenograft testing