Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brd9500 | 1630760-75-6 | 99.5% | C15H18FN3O2 | 10MG
SDP

Catalog No. 5000260976
Encompass_Preferred

BRD9500 is an orally active phosphodiesterase 3 (PDE3) inhibitor that selectively inhibits PDE3A and PDE3B at low-nanomolar concentrations and has demonstrated antitumor activity in assays that exploit PDE3A-SLFN12 interactions. It is supplied as a high-purity solid for use in biochemical and cell-based research.

  • Inhibits PDE3A and PDE3B with low-nanomolar potency.
  • Demonstrates antitumor activity via PDE3A-SLFN12 interactions.
  • Orally active small-molecule suitable for biochemical and cell-based assays.
  • Supplied as a solid powder with high purity.
  • Molecular formula C15H18FN3O2 and molecular weight 291.32 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; in solvent long-term at -80°C.

Catalog No. 50-002-60976 Supplier Medchemexpress LLC Supplier No. HY13635010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRD9500 is an orally active phosphodiesterase 3 (PDE3) inhibitor that selectively inhibits PDE3A and PDE3B at low-nanomolar concentrations and has demonstrated antitumor activity in assays that exploit PDE3A-SLFN12 interactions. It is supplied as a high-purity solid for use in biochemical and cell-based research.

  • Inhibits PDE3A and PDE3B with low-nanomolar potency.
  • Demonstrates antitumor activity via PDE3A-SLFN12 interactions.
  • Orally active small-molecule suitable for biochemical and cell-based assays.
  • Supplied as a solid powder with high purity.
  • Molecular formula C15H18FN3O2 and molecular weight 291.32 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; in solvent long-term at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.