Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brd9500 | 1630760-75-6 | MFCD32858286 | 99.5% | 291.32 g/mol | C15H18FN3O2 | 5 MG
SDP

Catalog No. 5000222112
Encompass_Preferred

Orally active small-molecule inhibitor of phosphodiesterase 3 (PDE3) with potent, low-nanomolar activity against PDE3A and PDE3B. Commonly used in biochemical and cellular studies to probe PDE3 function and has demonstrated antitumor effects in preclinical research.

  • Orally active PDE3 inhibitor.
  • PDE3A IC50: 10 nM; PDE3B IC50: 27 nM.
  • High purity (~99.5%).
  • Molecular weight 291.32 g/mol; formula C15H18FN3O2.
  • Available as solid and as 10 mM solution in DMSO.
  • Used to study cancer cell killing mediated by PDE3A/B interactions.

Catalog No. 50-002-22112 Supplier Medchemexpress LLC Supplier No. HY1363505MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orally active small-molecule inhibitor of phosphodiesterase 3 (PDE3) with potent, low-nanomolar activity against PDE3A and PDE3B. Commonly used in biochemical and cellular studies to probe PDE3 function and has demonstrated antitumor effects in preclinical research.

  • Orally active PDE3 inhibitor.
  • PDE3A IC50: 10 nM; PDE3B IC50: 27 nM.
  • High purity (~99.5%).
  • Molecular weight 291.32 g/mol; formula C15H18FN3O2.
  • Available as solid and as 10 mM solution in DMSO.
  • Used to study cancer cell killing mediated by PDE3A/B interactions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.