Learn More
Medchemexpress LLC HY-15647 5mg Medchemexpress, BRD9539 CAS:1374601-41-8 Purity:>98%
SDP
Catalog No. 502081764
Encompass_Preferred
Medchemexpress, HY-15647 5mg BRD9539 CAS:1374601-41-8 BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1764 Supplier Medchemexpress LLC Supplier No. HY156475MG
Medchemexpress, HY-15647 5mg BRD9539 CAS:1374601-41-8 BRD9539 is a histone methyltransferase G9a inhibitor with an IC50 of 6.3 μM. BRD9539 also inhibits PRC2 activity and is inactive against SUV39H1, NSD2 and DNMT1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.