Learn More
Cayman Chemical BRDT bromodomaIn 1humnre 100ug
SDP
Catalog No. 502952144
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 40.6 kDa • BRDT is similar to the RING3 protein family and possesses 2 bromodomain motifs and a PEST sequence motif, which is a region rich in proline (P), glutamic acid (E), serine (S), and threonine (T) residues known to have a short intracellular half-life.
Catalog No. 50-295-2144 Supplier Cayman Chemical Supplier No. 11548100 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 40.6 kDa • BRDT is similar to the RING3 protein family and possesses 2 bromodomain motifs and a PEST sequence motif, which is a region rich in proline (P), glutamic acid (E), serine (S), and threonine (T) residues known to have a short intracellular half-life.