Learn More
Cayman Chemical BRDT bromodomaIn 2humn rec
SDP
Catalog No. 502952523
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 41.2 kDa • BRDT shares homology with the RING3 protein. The two bromodomains of BRDT recognize acetylated histone H4. Loss of BRDT leads to defects in spermatogenesis. In addition to testis specific expression, BRDT was found in approximately 20% of non-small cell lung cancers.
Catalog No. 50-295-2523 Supplier Cayman Chemical Supplier No. 11649100 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • Mr: 41.2 kDa • BRDT shares homology with the RING3 protein. The two bromodomains of BRDT recognize acetylated histone H4. Loss of BRDT leads to defects in spermatogenesis. In addition to testis specific expression, BRDT was found in approximately 20% of non-small cell lung cancers.