Learn More
Cayman Chemical BRDT bromodomaIns 1 nd 2hu
SDP
Catalog No. 502971046
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 68.7 kDa
Catalog No. 50-297-1046 Supplier Cayman Chemical Supplier No. 14492100 UG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 68.7 kDa