Learn More
Biotium BrdU (5-Bromo-2'-Deoxyuridine)
Catalog No. 501964633
Encompass_Preferred
BrdU (5-Bromo-2′-deoxyuridine) can be incorporated into DNA during cell division and subsequently detected by a BrdU antibody. The probe is useful for studying cell-cycle kinetics. Properties: White solid soluble in DMSO; Store at -20⁰C; C9H11BrN2O5; MW: 307.10; [59-14-3].
Catalog No. 50-196-4633 Supplier Biotium Supplier No. 40024
BrdU (5-Bromo-2′-deoxyuridine) can be incorporated into DNA during cell division and subsequently detected by a BrdU antibody. The probe is useful for studying cell-cycle kinetics. Properties: White solid soluble in DMSO; Store at -20⁰C; C9H11BrN2O5; MW: 307.10; [59-14-3].