Learn More
Abcam BrdU (5-bromo-2'-deo x yuridine), Thymidine analog, 5G
Catalog No. 502583315
Encompass_Preferred
BrdU (5-bromo-2'-deoxyuridine), a thymidine analog, naturally incorporates into proliferating cells. Labels DNA, a useful marker for the study of DNA synthesis. Decreases TTP pools in vitro. Antitumor and radiation-sensitizing agent. Active in vitro and in vivo. - Cited in over 75 publications - Purity > 98% - Solubility: Dimethylacetamide ~ 200 (mg/mL), soluble in water to 50 mM
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3315 Supplier Abcam Supplier No. AB1425675G
BrdU (5-bromo-2'-deoxyuridine), a thymidine analog, naturally incorporates into proliferating cells. Labels DNA, a useful marker for the study of DNA synthesis. Decreases TTP pools in vitro. Antitumor and radiation-sensitizing agent. Active in vitro and in vivo. - Cited in over 75 publications - Purity > 98% - Solubility: Dimethylacetamide ~ 200 (mg/mL), soluble in water to 50 mM
The product is subject to the following: Abcam Restricted Use Statement