Learn More
Abcam BrdU Cell Proliferation ELISA Kit (colorimetric), 200TEST
Supplier:  Abcam AB126556200TEST
Encompass_Preferred
BrdU cell proliferation ELISA kit (colorimetric) is a sandwich ELISA used to quantify DNA synthesis and cell proliferation using BrdU with a sensitivity of 40 cells/well. - Rapid Protocol – assay completed in ~2.5 hours - Broad sample compatibility – works with adherent and suspension cells - Easy analysis with standard microplate readers - Cited in over 180 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0685878