Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ BrdU Kit for IHC/ICC Immunofluorescence eFluor™ 615

Catalog No. 8842659945
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-426-59945 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 88-426-59945 Supplier Invitrogen™ Supplier No. 8842659945
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide and formaldehyde

Includes

  • BrdU Fixation Buffer
  • BrdU Antigen Retrieval Solution (10X)
  • BrdU Blocking Solution
  • Anti-BrdU Biotin (clone BU20A) : 10 μg
  • Streptavidin for BrdU IHC/ICC eFluor 615 : 3 μg

Contains reagents and buffers to identify and examine proliferating cells by immunohistochemical or immunocytochemical analysis. Optimized for IHC with both frozen and formalin-fixed paraffin-embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.

Cells are labeled in vitro or in vivo with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine, which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle. Following fixation and antigen retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using a streptavidin-conjugated fluorophore.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate BrdU
Detection Method Fluorometric
Quantity 50 tests
Product Line eBioscience
For Use With (Application) ICC (Immunochemistry), IHC (Immunohistochemistry)
Product Type IHC/ICC BrdU Kit
Target Species All species

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.