Learn More
Enzo Life Sciences BrdU monoclonal antibody (2B1) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIMSA200E
Encompass_Preferred
Clone: 2B1. Host: Mouse. Isotype: IgG1. Immunogen: Iododeoxyuridine-Ovalbumin. Species reactivity: Species independent. Applications: Flow Cytometry, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7350