Learn More
Sparkfun Electronics BREADBOARD - FULL-SIZE (BARE)
Catalog No. NC1071022
Encompass_Preferred
Breadboard - Full-Size (Bare). 6.5 x 2.1 x 0.38" (165.1 x 54.29 x 9.68mm). This is your tried and true full size solderless breadboard! It has 2 split power buses, 10 columns, and 63 rows - with a total of 830 tie in points. All pins are spaced by a standard 0.1". The two sets of five columns are separated by about 0.3", perfect for straddling a DIP package over. The board accepts wire sizes in the range of 20-29AWG.
Catalog No. NC1071022 Supplier Sparkfun Electronics Supplier No. PRT12615
Breadboard - Full-Size (Bare). 6.5 x 2.1 x 0.38" (165.1 x 54.29 x 9.68mm). This is your tried and true full size solderless breadboard! It has 2 split power buses, 10 columns, and 63 rows - with a total of 830 tie in points. All pins are spaced by a standard 0.1". The two sets of five columns are separated by about 0.3", perfect for straddling a DIP package over. The board accepts wire sizes in the range of 20-29AWG.