Learn More
Electron Microscopy Sciences Break-Safe™ Break-Safe Ampoule Opener
SDP
Catalog No. 50366003
Encompass_Preferred
Box style design allows one handed opening of up to 3 ampules at a time with unobstructed viewing of ampule contents at all time. It in constructed of copolyester plastic which has a broad chemical resistance. Gently pressing down on the lid snaps ampule necks clean while keeping them contained within the device and away from fingers. After removing ampule contents, simply turn the device upside down over a trash receptacle to safely discard ampules and tops.
The item includes 4 inserts to allow opening of 1ml, 2ml, 5ml and 10 ml ampules.
Catalog No. 50-366-003 Supplier Electron Microscopy Sciences Supplier No. 60607
Box style design allows one handed opening of up to 3 ampules at a time with unobstructed viewing of ampule contents at all time. It in constructed of copolyester plastic which has a broad chemical resistance. Gently pressing down on the lid snaps ampule necks clean while keeping them contained within the device and away from fingers. After removing ampule contents, simply turn the device upside down over a trash receptacle to safely discard ampules and tops.
The item includes 4 inserts to allow opening of 1ml, 2ml, 5ml and 10 ml ampules.