Learn More
MSC Diversey Oven And Grill Cleaner, Aerosol, 19 oz, Ready to Use
Catalog No. 502839143
Encompass
This institutional formula cleaner provides heavy-duty cleaning power for fast and easy cleaning of a variety of institutional, industrial, and commercial surfaces. It works immediately on warm or cold ovens and effectively cleans materials such as stainless steel, porcelain enamel, iron, undecorated ceramic tile, concrete, and glass. The aerosol spray conveniently reaches tough-to-reach areas.
  • Ready-to-use liquid formula with no scent.
  • NSF listed and Kosher certified.
  • Breaks down tough, baked-on grease and grime on contact, even on cold surfaces.
  • Minimizes scrubbing and scouring on commercial ovens, grills, and fryers.
  • Effectively cleans surfaces including stainless steel, porcelain enamel, iron, undecorated ceramic tile, concrete, and glass.
  • Conveniently sprays from any angle to penetrate tough-to-reach areas.
Catalog No. 50-283-9143 Supplier MSC Supplier No. 50528256
This institutional formula cleaner provides heavy-duty cleaning power for fast and easy cleaning of a variety of institutional, industrial, and commercial surfaces. It works immediately on warm or cold ovens and effectively cleans materials such as stainless steel, porcelain enamel, iron, undecorated ceramic tile, concrete, and glass. The aerosol spray conveniently reaches tough-to-reach areas.
  • Ready-to-use liquid formula with no scent.
  • NSF listed and Kosher certified.
  • Breaks down tough, baked-on grease and grime on contact, even on cold surfaces.
  • Minimizes scrubbing and scouring on commercial ovens, grills, and fryers.
  • Effectively cleans surfaces including stainless steel, porcelain enamel, iron, undecorated ceramic tile, concrete, and glass.
  • Conveniently sprays from any angle to penetrate tough-to-reach areas.