Learn More
Fine Science Tools Scalpel Blades-Breakable, 10/pk
SDP
Catalog No. NC9827009
Encompass
Product: Scalpel Blades - Breakable/Variable Cutting Edge/ Blade Shape: Straight/ Cutting Edge: Variable/ Thickness: 0.1mm/ Alloy / Material: Carbon Steel. These come individually wrapped and are the only blades that break cleanly. The alloy has a high carbon content which provides a longer lasting cutting edge. Not autoclavable. 10/pk
Catalog No. NC9827009 Supplier Fine Science Tools Supplier No. 1005000
Product: Scalpel Blades - Breakable/Variable Cutting Edge/ Blade Shape: Straight/ Cutting Edge: Variable/ Thickness: 0.1mm/ Alloy / Material: Carbon Steel. These come individually wrapped and are the only blades that break cleanly. The alloy has a high carbon content which provides a longer lasting cutting edge. Not autoclavable. 10/pk