Learn More
Electron Microscopy Sciences CoreDish™ Multiple Biopsy Containers Breast, 5 compartments
SDP
Catalog No. 501901136
Encompass_Preferred
Multi-compartment containersin the shape of a dish are half prefilled with 10% neutral buffered Formalin and used for holding and transporting biopsies. It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. They measure only 15 mm H x 95 mm diameter. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted.
No longer necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion. A label allows recording the related information, patient ID, doctor, date and time.
Catalog No. 50-190-1136 Supplier Electron Microscopy Sciences Supplier No. 1573610
Multi-compartment containersin the shape of a dish are half prefilled with 10% neutral buffered Formalin and used for holding and transporting biopsies. It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. They measure only 15 mm H x 95 mm diameter. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted.
No longer necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion. A label allows recording the related information, patient ID, doctor, date and time.