Learn More
Discovery Life Sciences Breast Ca Act Tx IO PBMC
SDP
Catalog No. 5000804024
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04024 Supplier Discovery Life Sciences Supplier No. 120940
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays