Learn More
Discovery Life Sciences Breast Ca Prog/Ref/Rel PBMC
SDP
Catalog No. 5000804056
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04056 Supplier Discovery Life Sciences Supplier No. 120960
Add to Cart
Add to Cart
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays