Learn More
US Biomax Inc Breast cancer tissue array, 75 cases/150 cores, replacing BR1505d
SDP
Catalog No. NC3237344 Shop All US Biomax Inc Products
Encompass
Breast cancer tissue array, including pathology grade, TNM, clinical stage, and IHC(HER2/ER/PR/Ki67), 75 cases/150 cores, replacing BR1505d
Catalog No. NC3237344 Supplier US Biomax Inc Supplier No. BR1505E
Breast cancer tissue array, including pathology grade, TNM, clinical stage, and IHC(HER2/ER/PR/Ki67), 75 cases/150 cores, replacing BR1505d