Learn More
StatLab Breast Carcinoma c-erB-2PLUS Control Slides
SDP
Catalog No. 22024916
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22-024-916 25 Pk.
1 options
Catalog No. 22-024-916 Supplier StatLab Supplier No. CSB0725P
Add to Cart
Add to Cart
Use breast carcinoma c-erB-2PLUS control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.