Learn More
StatLab Breast Carcinoma IHC Control Slides
SDP
Catalog No. 21033323
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
21-033-323 25 Pk.
1 options
Catalog No. 21-033-323 Supplier StatLab Supplier No. CSB0825P
Add to Cart
Add to Cart
Use breast carcinoma IHC control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.