Learn More
INNOVEX BIOSCIENCES BREAST PANEL MULTI-TUMOR CONTROL SLIDES 25
SDP
Catalog No. 50486691
Encompass_Preferred
Breast panel Multi Tumor control slide; Each slide contains 5 pieces of 4 micron sections of breast tumor tissues each positive for ER, PR, ki-67, BRST-2, E-Cadherin and Cytokeratins for sentinel nodes; 25 slides per package
Catalog No. 50-486-691 Supplier INNOVEX BIOSCIENCES Supplier No. TS90125
Breast panel Multi Tumor control slide; Each slide contains 5 pieces of 4 micron sections of breast tumor tissues each positive for ER, PR, ki-67, BRST-2, E-Cadherin and Cytokeratins for sentinel nodes; 25 slides per package