Learn More
Andwin Scientific BREATH-EASIER MEMBRANES 100/PK
SDP
Catalog No. 50930378
Encompass_Preferred
The Breathe-EASIER Membranes can be utilized for efficient cell growth and freeze drying. This product provides a consistent gas exchange. The membranes do not bind in centrifuge rotors. These are pre-cut, sterile, and free of cytotoxins. It is sold in a pack of 100.
Catalog No. 50-930-378 Supplier Andwin Scientific Supplier No. 7053720
The Breathe-EASIER Membranes can be utilized for efficient cell growth and freeze drying. This product provides a consistent gas exchange. The membranes do not bind in centrifuge rotors. These are pre-cut, sterile, and free of cytotoxins. It is sold in a pack of 100.