Learn More
Andwin Scientific BREATH EASY 100/PK
SDP
Catalog No. 50930375
Encompass_Preferred
Made with polyurethane, Breathe-Easy is an adhesive that is ideal for use in cell culture applications. It is able to work within the temperature range of -80C to 100C. This product has a thickness of 1 mil (0.001 in.). Breathe-Easy is gas-permeable and is UV Transparent to 300nm. It is sold in a pack of 100.
Catalog No. 50-930-375 Supplier Andwin Scientific Supplier No. 7053610
Made with polyurethane, Breathe-Easy is an adhesive that is ideal for use in cell culture applications. It is able to work within the temperature range of -80C to 100C. This product has a thickness of 1 mil (0.001 in.). Breathe-Easy is gas-permeable and is UV Transparent to 300nm. It is sold in a pack of 100.