Learn More
USA Scientific Inc Breathe-Easy film, sterile
Catalog No. NC9520988
Encompass
Breathe-Easy polyurethane film, -80C to +100C, sterile, 100/box
Catalog No. NC9520988 Supplier USA Scientific Inc Supplier No. 91236100
Breathe-Easy polyurethane film, -80C to +100C, sterile, 100/box