Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tian's International Inc. Breathable Cleanroom Hood

Catalog No. 19098326 Shop All Tians International Products
Change view
Click to view available options
Material:
Spunbond Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-098-326 Spunbond Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-098-326 Supplier Tian's International Inc. Supplier No. 416550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tian's International, Inc. Breathable Cleanroom Hood for use in critical and low particulate clean environments.

  • Low lint
  • Full coverage
  • Elastic face opening can be worn with a full face, or partial opening
  • Elastic neck closure
  • Non-woven, cloth-like fabric is comfortable to wear
  • Individually inspected for sizing, stitching, and workmanship

Specifications

Product Type Cleanroom Hood
Size Universal
Material Spunbond Polypropylene
Color White
Closure Type Elastic
Disposable Single-use
Latex Free Yes
Model 416550
Seam Type Serged
Sterility Non-sterile
Packaging Type Cleanroom Packaged
Quantity 500/Cs.
Width (English) 13 in.
Height (Metric) 42 cm
Diameter (Metric) 34 cm
For Use With (Application) Critical Environment
Hood Type Open Face
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.