Learn More
Bellco Glass, Inc. Breathable Sealing
SDP
Catalog No. 502329339
Encompass_Preferred
Breathable Sealing Film promotes uniform gas exchange and can be pierced easily with pipet tipsReduces outer well evaporation and cross contaminationRecommended for use between -20°C and 120°C Pack Description50/Re-sealable BagCase Quantity50SterileYesPrimary ColorNaturalPrimary MaterialFilm: RayonSecondary MaterialAdhesive: Acrylic
Catalog No. 50-232-9339 Supplier Bellco Glass, Inc. Supplier No. 229130000
Breathable Sealing Film promotes uniform gas exchange and can be pierced easily with pipet tipsReduces outer well evaporation and cross contaminationRecommended for use between -20°C and 120°C Pack Description50/Re-sealable BagCase Quantity50SterileYesPrimary ColorNaturalPrimary MaterialFilm: RayonSecondary MaterialAdhesive: Acrylic