Learn More
USA Scientific Inc Breathe-Easy tube membranes
Catalog No. NC9623767
Encompass
Breathe-Easy transparent gas exchange membranes for microcentrifuge tubes, sterile, 100/pack
Catalog No. NC9623767 Supplier USA Scientific Inc Supplier No. 91261000
Breathe-Easy transparent gas exchange membranes for microcentrifuge tubes, sterile, 100/pack