Learn More
Andwin Scientific Breathe-Easier Sealing Film 100/pk
SDP
Catalog No. NC1664397
Encompass_Preferred
Breathe-Easier sealing film allows rapid and uniform transmission of oxygen, carbon dioxide and water vapors to stimulate cell growth.
Catalog No. NC1664397 Supplier Andwin Scientific Supplier No. BERM2000
Breathe-Easier sealing film allows rapid and uniform transmission of oxygen, carbon dioxide and water vapors to stimulate cell growth.