Learn More
Sigma Aldrich Fine Chemicals Biosciences Breathe Easier sealing mem
Catalog No. 502901802
Encompass
Gas-Permeable Sealing Membrane - A fresh Breath for Cell Growth TechnologyBreathe-Easier sealing membrane offers an exceedingly high rate of gas exchange to allow maximum cell growth rate. When adhered to a microtiter plate the 5.5 mil thick membrane insures a uniform rate of gas exchange for all wells while sustaining a contaminant-free environment within the well. Conventional microtiter lids disrupt uniform gas exchange from well to well by favoring those wells along the perimeter of the plate. With Breathe-Easier sterility is achieved using gamma irradiation. Unlike ethylene oxide sterilization this sterilization method assures that no cytotoxins are introduced into the membranes. Breathe Easier will not inhibit fibroblast cell growth or division and the membrane is fDA approved for blood and cell contact in wond healing. The acrylic adhesive exhibits excellent bonding characteristics with all different plastic microtiter plates. High Gas Permeability- Rapid and uniform t
Catalog No. 50-290-1802 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z763624100EA
Gas-Permeable Sealing Membrane - A fresh Breath for Cell Growth TechnologyBreathe-Easier sealing membrane offers an exceedingly high rate of gas exchange to allow maximum cell growth rate. When adhered to a microtiter plate the 5.5 mil thick membrane insures a uniform rate of gas exchange for all wells while sustaining a contaminant-free environment within the well. Conventional microtiter lids disrupt uniform gas exchange from well to well by favoring those wells along the perimeter of the plate. With Breathe-Easier sterility is achieved using gamma irradiation. Unlike ethylene oxide sterilization this sterilization method assures that no cytotoxins are introduced into the membranes. Breathe Easier will not inhibit fibroblast cell growth or division and the membrane is fDA approved for blood and cell contact in wond healing. The acrylic adhesive exhibits excellent bonding characteristics with all different plastic microtiter plates. High Gas Permeability- Rapid and uniform t