Learn More
Andwin Scientific BREATHE-EASY ADH FILM G,100/PK
SDP
Catalog No. 50820936
Encompass_Preferred
The 25 �m Breathe-Easy film can work under the temperature range of -80�C to 100�C. With the ability to absorb oxygen, carbon dioxide, and water vapors, this product is an ideal sealing film. It is also UV transparent down to 300 nm, non-cytotoxic, and sterile. A brayer or sealing paddle is needed to perform a proper sealing. This product is sold in a pack of 100.
Catalog No. 50-820-936 Supplier Andwin Scientific Supplier No. 91236100
The 25 �m Breathe-Easy film can work under the temperature range of -80�C to 100�C. With the ability to absorb oxygen, carbon dioxide, and water vapors, this product is an ideal sealing film. It is also UV transparent down to 300 nm, non-cytotoxic, and sterile. A brayer or sealing paddle is needed to perform a proper sealing. This product is sold in a pack of 100.