Learn More
3M™ Breathe Easy™ Battery Adapter Upgrade Kit
Catalog No. 19070838 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-070-838 1 Ea.
1 options
Catalog No. 19-070-838 Supplier 3M™ Supplier No. 7100144938
Add to Cart
Add to Cart
Battery adapter upgrade kit allows use of the TR-630 battery with First Responder Breathe Easy™ PAPR systems.
  • Easy to use
  • Easy to clean
  • Works with 3M™ Standard Battery TR-630 providing longer run time than the 3M™ Breathe Easy™ Turbo Powered Air Purifying Respirator (PAPR) NiMH Battery Pack BP-15
  • Comfortable
  • Replaces BP-15 Battery
  • 3M Catalog# RBE-600
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 5.5 to 13 hr.
Battery Type High Capacity Lithium Ion
Certifications/Compliance NIOSH Approved
Product Type Battery Attachment Tool
Includes Battery adapter (TR-659), battery (TR-630), charger (TR-641N) and easy clean holster (TR-657)
Model RBE-600
Quantity 1 Ea.