Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Breathe Easy™ High Efficiency Cartridge

Catalog No. 19070835 Shop All 3M Products
Change view
Click to view available options
Quantity: 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-070-835 Quantity: 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-835 Supplier 3M™ Supplier No. 50051138729177

This NIOSH-approved cartridge helps provide high efficiency respiratory protection in environments containing toxic industrial chemicals. Use with the RRPAS 6000 Series facepieces or BE-10 Series hood powered supplied air respirator systems.

  • Helps provide high efficiency respiratory protection in environments containing toxic industrial chemicals
  • Respiratory protection against: organic vapor, acid gas (sulfur dioxide, chlorine and hydrogen chloride), ammonia/methylamine, hydrogen fluoride, chlorine dioxide, and formaldehyde
  • Can also filter a wide range of chemical warfare agents such as: nerve, mustard, tear and blood agents; chlorine, phosgene, chloropicrin, and diphenylchloroarsine
  • 3M Catalog# 453-03-02R06

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Acid Gas, Organic Vapor
Description Multi gas and vapor cartridge with high efficiency (HE) particulate protection
Color Magenta/Olive
For Use With (Equipment) RPAS 6000 Series facepieces or BE-10 Series hood powered supplied air respirator systems
Model 453-03-02R06
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.