Learn More
Electron Microscopy Sciences Micro-permeable Breathe-EASY Breathable Tube Membranes
SDP
Catalog No. 501899350
Encompass_Preferred
Micro-permeable membranes for microtubes ideal for cell growth and freeze drying. Permits consistent gas exhange. Easy microtube cap closure. Will not bind in centrifuge rotors. Easy-to-use finger tab for quick liner removal. Pre-cut to fit a variety of popular tube sizes. Sterile and free of cytotoxins.
Catalog No. 50-189-9350 Supplier Electron Microscopy Sciences Supplier No. 7053710
Micro-permeable membranes for microtubes ideal for cell growth and freeze drying. Permits consistent gas exhange. Easy microtube cap closure. Will not bind in centrifuge rotors. Easy-to-use finger tab for quick liner removal. Pre-cut to fit a variety of popular tube sizes. Sterile and free of cytotoxins.