Learn More
Sigma Aldrich Fine Chemicals Biosciences Breathe-Easy sealing membrane, 1pk
Catalog No. NC0674973
Encompass
Synonym: breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture. Medical grade polyurethane membrane with FDA-approved acrylic adhesive pre-cut to fit standard multiwell plates. The membrane is permeable to oxygen, carbon dioxide and water vapor, creating separate, highly uniform microculture vessels from a multi-well plate.
Catalog No. NC0674973 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3800591PAK
Synonym: breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture. Medical grade polyurethane membrane with FDA-approved acrylic adhesive pre-cut to fit standard multiwell plates. The membrane is permeable to oxygen, carbon dioxide and water vapor, creating separate, highly uniform microculture vessels from a multi-well plate.