Learn More
Med Vet International Breathe Safe Respiration Monitor, J0166RM
Catalog No. 5000583910
Encompass_Preferred
This tiny monitor makes a great apnea alert monitor. The small microprocessor beeps with every breath with a very high sensitivity.Easy to use: just connect between endotracheal tube and anesthesia circuitNo adjustments: a sophisticated algorithm recognizesand indicates every breath with a beep, workingfrom a 1 lb. kitten to a 200 lb. St. Bernard.Apnea alert; a distinctive alert sounds if patient fails tobreathe for 45 secondsAuto shut-offA high density long-life lithium battery provides years of serviceOne-year warranty
Catalog No. 50-005-83910 Supplier Med Vet International Supplier No. JOR166RM
This tiny monitor makes a great apnea alert monitor. The small microprocessor beeps with every breath with a very high sensitivity.Easy to use: just connect between endotracheal tube and anesthesia circuitNo adjustments: a sophisticated algorithm recognizesand indicates every breath with a beep, workingfrom a 1 lb. kitten to a 200 lb. St. Bernard.Apnea alert; a distinctive alert sounds if patient fails tobreathe for 45 secondsAuto shut-offA high density long-life lithium battery provides years of serviceOne-year warranty