Learn More
Research Products International Corp Breathe-Easy Sealing Membrane, Regular Strength, 5 x 3 1/4 Inch Rectangles, 100 per Package
SDP
Catalog No. 50550304
Encompass
Breathe-Easy provides a gas permeable seal for microwell plates. Plates covered with Breathe-Easy allow a uniform rate of oxygen and CO2 exchange across the membrane. Promotes contaminant free growth of cell cultures, cellular based biological and immunological assays, cell selection and cell screening.
Plates covered with Breathe-Easy membrane can be inverted and agitated without cross contamination samples. Adhesive is FDA approved and will not interfere with human fibroblast cell growth and division. Resistant to most organic and caustic agents and withstands temperature from -80°C to 100°C.
Easy to use. Remove release liner, roll brayer several times in all directions to loosen protective top layer and peel away top layer.
Removable, not reusable.
Catalog No. 50-550-304 Supplier Research Products International Corp Supplier No. 248738
Breathe-Easy provides a gas permeable seal for microwell plates. Plates covered with Breathe-Easy allow a uniform rate of oxygen and CO2 exchange across the membrane. Promotes contaminant free growth of cell cultures, cellular based biological and immunological assays, cell selection and cell screening.
Plates covered with Breathe-Easy membrane can be inverted and agitated without cross contamination samples. Adhesive is FDA approved and will not interfere with human fibroblast cell growth and division. Resistant to most organic and caustic agents and withstands temperature from -80°C to 100°C.
Easy to use. Remove release liner, roll brayer several times in all directions to loosen protective top layer and peel away top layer.
Removable, not reusable.