Learn More
Research Products International Corp Breathe-Easy Petri Dish Strip Seals, 100 per Pack
SDP
Catalog No. 501367706
Encompass
Apply Breathe-Easy strip seals to guard against contamination during culture incubation or long term storage of petri dishes. Strips are pre-cut from gas permeable membrane that allows the exchange of oxygen and CO2 for proper cell growth. Sealed dishes can be incubated or stored at temperatures from -80°C to +100°C.
Pre-cut measured strips wrap completely around a 100 mm diameter petri dish.
Catalog No. 50-136-7706 Supplier Research Products International Corp Supplier No. 247124
Apply Breathe-Easy strip seals to guard against contamination during culture incubation or long term storage of petri dishes. Strips are pre-cut from gas permeable membrane that allows the exchange of oxygen and CO2 for proper cell growth. Sealed dishes can be incubated or stored at temperatures from -80°C to +100°C.
Pre-cut measured strips wrap completely around a 100 mm diameter petri dish.