Learn More
USA Scientific Inc Breathe-Easier film, sterile
Catalog No. NC1758550
Encompass
Breathe-Easier non-woven rayon film, 5.5 mil thick, -20C to +80C, sterile, 100/box
Catalog No. NC1758550 Supplier USA Scientific Inc Supplier No. 91262100
Breathe-Easier non-woven rayon film, 5.5 mil thick, -20C to +80C, sterile, 100/box