Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ BreathID™ HP Smart Refurbished

Catalog No. 23655114 Shop All Meridian Bioscience Products
Change view
Click to view available options
Clia Complexity:
Waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
23-655-114 Waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-655-114 Supplier Meridian Bioscience™ Supplier No. VS07000R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BreathID™ Smart is a urea breath test platform for the detection of H. pylori and confirmation of eradication.

The state-of-the-art BreathID™ Smart urea breath test platform for H. pylori detection intuitively combines market-leading diagnostic accuracy with efficient automation, all in a compact footprint for maximizing mid-range testing throughput. Whether for initial active infection detection or confirmation of eradication, Meridian is your trusted partner in gastrointestinal diagnostics.

  • Perfect platform for low to mid-volume laboratories and clinics
    • Test samples have a two-week window for analysis and require no special storage
    • 4 test samples per auto sampler
  • Under 10 minutes for full batch analysis, 2 minutes per test
  • User-friendly interface
    • Touchscreen for intuitive process management
    • Scannable barcodes maximize speed of data entry
    • Automated QC selection and label generation
  • Flexible data management options
    • Print, export, or transmit results
    • Secure, seamless integration with LIS
  • Minimizes potential errors before they happen
    • Automatic identification and correction of reversed bags
    • Coordinated indicator signals guide batch assembly and bag placement
    • Scannable barcodes minimize human error

Specifications

CE Marker Yes
Clia Complexity Waived
Detectable Analytes H. pylori
Format Kit
Sample Type 4 Test samples per auto sampler
Sensitivity 100%
Type Breath Test System
Disposable Reusable
Specificity 97.90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.