Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ BreathID™ IDkit Hp™ Two
GSA_VA

Catalog No. 23655115 Shop All Meridian Bioscience Products
Change view
Click to view available options
Clia Complexity:
Waived
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
23-655-115 Waived
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-655-115 Supplier Meridian Bioscience™ Supplier No. AC00063
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A non-invasive urea breath test for the detection of H. pylori and confirmation of eradication.

Non-invasive specimen collection allows for improved patient satisfaction and compliance.

  • For use with the BreathID Hp Lab and BreathID™ Smart Systems.
  • Kit Components: Blue BASELINE breath bag, grey POST-INGESTION breath bag, 75 mg 13C-urea tablet, 4.3 g package of Citrica (citric acid), straw drinking cup, user guide, 4 labels, sample transport bag, and package insert

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

CE Marker Yes
Clia Complexity Waived
Detectable Analytes H. pylori
Format Kit
Sample Type Breath
Type Urea Breath Test
For Use With (Equipment) BreathID Hp Lab and BreathID Smart Systems
Includes Blue BASELINE breath bag, gray POST-INGESTION breath bag, 75 mg 13C-urea tablet, 4.3 g package of Citrica (citric acid), straw drinking cup, user guide, 4 labels, sample transport bag, and package Insert
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.