Learn More
3M™ Breathing Room Assembly
Catalog No. 1798596R Shop All 3M Products
Change view
Click to view available options
Quantity:
5/Cs.
Catalog No. Quantity
17-985-96R 5/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-96R Supplier 3M™ Supplier No. 7000126023
This item is not returnable. View return policy
For use as a replacement part for 7000 Series full facepieces. Does not include Speaking Diaphragm Kit 7895.
  • Approved 3M replacement part
  • Designed for 3M Full Facepieces 7800 Series (sold separately)
  • Replacement part can extend service life of facepiece
  • Use can potentially result in respiratory equipment cost savings
  • Assembly maintains original service performance of facepiece
TRUSTED_SUSTAINABILITY
Specifications
Product Type Replacement Parts and Accessories
For Use With (Application) Assembly and Mechanical, Painting, Cleaning, Facility Maintenance
For Use With (Equipment) 3M™ Full Facepiece 7800 Series
Model 7916-5
Quantity 5/Cs.